missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluoroisopropyl Methyl Ether 98.0+%, TCI America™
Supplier: TCI America H15245G
Specifications
| Hexafluoroisopropyl Methyl Ether | |
| Colorless | |
| C4H4F6O | |
| 5 g | |
| 1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Isoindoklon | |
| COC(C(F)(F)F)C(F)(F)F | |
| 182.065 | |
| 182.07 | |
| Liquid |
| 13171-18-1 | |
| 50°C | |
| MFCD00221772 | |
| 3271 | |
| VNXYDFNVQBICRO-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoro-2-methoxypropane | |
| 25749 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 13171-18-1 | |
| 182.065 | |
| VNXYDFNVQBICRO-UHFFFAOYSA-N | |
| 25749 | |
| COC(C(F)(F)F)C(F)(F)F |
| C4H4F6O | |
| MFCD00221772 | |
| 1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Isoindoklon | |
| 1,1,1,3,3,3-hexafluoro-2-methoxypropane |
Safety and Handling
EINECSNumber : (2)-4050
RTECSNumber : KO2282000
TSCA : No