missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluoroacetylacetone 95.0+%, TCI America™
$96.94 - $283.33
Chemical Identifiers
| CAS | 1522-22-1 |
|---|---|
| Molecular Formula | C5H2F6O2 |
| Molecular Weight (g/mol) | 208.059 |
| MDL Number | MFCD00000426 |
| InChI Key | QAMFBRUWYYMMGJ-UHFFFAOYSA-N |
| Synonym | hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane |
| PubChem CID | 73706 |
| IUPAC Name | 1,1,1,5,5,5-hexafluoropentane-2,4-dione |
| SMILES | C(C(=O)C(F)(F)F)C(=O)C(F)(F)F |
Chemical Identifiers
| 1522-22-1 | |
| 208.059 | |
| QAMFBRUWYYMMGJ-UHFFFAOYSA-N | |
| 73706 | |
| C(C(=O)C(F)(F)F)C(=O)C(F)(F)F |
| C5H2F6O2 | |
| MFCD00000426 | |
| hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane | |
| 1,1,1,5,5,5-hexafluoropentane-2,4-dione |
Specifications
| 1522-22-1 | |
| 70°C | |
| MFCD00000426 | |
| 2920 | |
| QAMFBRUWYYMMGJ-UHFFFAOYSA-N | |
| 1,1,1,5,5,5-hexafluoropentane-2,4-dione | |
| 73706 | |
| ≥95.0% (GC) | |
| Hexafluoroacetylacetone |
| Yellow | |
| C5H2F6O2 | |
| 5 g | |
| hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane | |
| C(C(=O)C(F)(F)F)C(=O)C(F)(F)F | |
| 208.059 | |
| 208.06 | |
| Liquid |
Safety and Handling
TSCA : Yes