missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol 98.0+%, TCI America™

Catalog No. p-7130956
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12795G 5 g
H127925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12795G Supplier TCI America Supplier No. H12795G
Only null left

Chemical Identifiers

CAS 918-21-8
Molecular Formula C6H2F12O2
Molecular Weight (g/mol) 334.06
MDL Number MFCD00427700
InChI Key GKDCWKGUOZVDFX-UHFFFAOYSA-N
Synonym Perfluoropinacol
PubChem CID 13518
IUPAC Name 1,1,1,4,4,4-hexafluoro-2,3-bis(trifluoromethyl)butane-2,3-diol
SMILES OC(C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 26°C
Color White-Yellow
Boiling Point 129°C
Quantity 5 g
Formula Weight 334.06
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol
RTECSNumber EK0650000
TSCA No