missing translation for 'onlineSavingsMsg'
Learn More

Hexaethylene Glycol 98.0+%, TCI America™

Catalog No. p-7131041
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14325G 5 g
H143225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14325G Supplier TCI America Supplier No. H14325G
Only null left

Chemical Identifiers

CAS 2615-15-8
Molecular Formula C12H26O7
Molecular Weight (g/mol) 282.333
MDL Number MFCD00002877
InChI Key IIRDTKBZINWQAW-UHFFFAOYSA-N
Synonym hexaethylene glycol, hexagol, 3,6,9,12,15-pentaoxaheptadecane-1,17-diol, hexaoxyethylene glycol, peg-6, polyethylene glycol 300, polyoxyethylene 6, hexaethyleneglycol, 2-2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethoxy ethanol, ho-peg6-oh
PubChem CID 17472
ChEBI CHEBI:49793
IUPAC Name 2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCOCCO)O

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 217°C
Quantity 5 g
Formula Weight 282.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexaethylene Glycol
EINECSNumber (2)-0441
RTECSNumber MM3670000
TSCA No