missing translation for 'onlineSavingsMsg'
Learn More

Hexaethyl phosphorous triamide, 97%

Catalog No. AAA1249006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1249006 5 g
AAA1249014 25 g
AAA1249022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1249006 Supplier Thermo Scientific Chemicals Supplier No. A1249006

CAS: 2283-11-6 | C12H30N3P | 247.37 g/mol

Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.

Chemical Identifiers

CAS 2283-11-6
MDL Number MFCD00009041

Specifications

Density 0.903
Boiling Point 245°C to 246°C
Flash Point 58°C (136°F)
Molecular Formula C8H4F3NO
Refractive Index 1.4737
Quantity 5 g
UN Number UN1993
Beilstein 636187
Sensitivity Air Sensitive
Synonym 2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
Solubility Information Insoluble in water.
InChI Key GZWGTVZRRFPVAS-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=O
IUPAC Name 1-isocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 187.12
PubChem CID 16794
Formula Weight 247.37
Percent Purity 97%
Chemical Name or Material Hexaethyl phosphorous triamide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 218-920-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only