missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Perchlorate 98.0+%, TCI America™

Catalog No. H07351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07351G 1 g
H07355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07351G Supplier TCI America Supplier No. H07351G

Chemical Identifiers

CAS 6941-37-3
Molecular Formula C19H42ClNO4
Molecular Weight (g/mol) 384.00
MDL Number MFCD00144895
InChI Key KXNZYPBRSATHKV-UHFFFAOYSA-M
Synonym Cetyltrimethylammonium Perchlorate
PubChem CID 15520692
IUPAC Name hexadecyltrimethylazanium perchlorate
SMILES [O-][Cl](=O)(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Quantity 1 g
UN Number 1479
Formula Weight 384.00
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyltrimethylammonium Perchlorate
EINECSNumber (2)-0184&(9)-0795&(9)-1971
TSCA No