missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyltrimethoxysilane 85.0+%, TCI America™
Supplier: TCI America H137625G
Specifications
| Hexadecyltrimethoxysilane | |
| Colorless | |
| C19H42O3Si | |
| 25 g | |
| RSKGMYDENCAJEN-UHFFFAOYSA-N | |
| hexadecyltrimethoxysilane | |
| 85406 | |
| ≥85.0% (GC) |
| 16415-12-6 | |
| 155°C | |
| MFCD00069149 | |
| Trimethoxy(hexadecyl)silane | |
| CCCCCCCCCCCCCCCC[Si](OC)(OC)OC | |
| 346.63 | |
| 346.63 | |
| Liquid |
Chemical Identifiers
| 16415-12-6 | |
| 346.63 | |
| RSKGMYDENCAJEN-UHFFFAOYSA-N | |
| 85406 | |
| CCCCCCCCCCCCCCCC[Si](OC)(OC)OC |
| C19H42O3Si | |
| MFCD00069149 | |
| Trimethoxy(hexadecyl)silane | |
| hexadecyltrimethoxysilane |
Safety and Handling
TSCA : Yes