missing translation for 'onlineSavingsMsg'
Learn More

Hexadecylpyridinium Chloride Monohydrate 98.0+%, TCI America™

Catalog No. p-7130321
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H007825G 25 g
H0078100G 100 g
H0078500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H007825G Supplier TCI America Supplier No. H007825G
Only null left

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.01
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
Synonym cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
PubChem CID 22324
ChEBI CHEBI:3566
IUPAC Name 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

Specifications

Melting Point 84°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 339.99
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecylpyridinium Chloride Monohydrate
RTECSNumber UU5075000
TSCA Yes