missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecylpyridinium Chloride Monohydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 84°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 2811 |
| Formula Weight | 339.99 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Hexadecylpyridinium Chloride Monohydrate |
Safety and Handling
| RTECSNumber | UU5075000 |
| TSCA | Yes |