missing translation for 'onlineSavingsMsg'
Learn More

Hexadecylphosphonic Acid 98.0+%, TCI America™

Catalog No. H14885G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14885G 5 g
H14881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14885G Supplier TCI America Supplier No. H14885G
Only null left

Chemical Identifiers

CAS 4721-17-9
Molecular Formula C16H35O3P
Molecular Weight (g/mol) 306.427
MDL Number MFCD00015836
InChI Key JDPSFRXPDJVJMV-UHFFFAOYSA-N
PubChem CID 13783525
IUPAC Name hexadecylphosphonic acid
SMILES CCCCCCCCCCCCCCCCP(=O)(O)O

Specifications

Melting Point 95°C
Color White
Quantity 5 g
Formula Weight 306.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecylphosphonic Acid