missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyl Lactate 75.0+%, TCI America™

Catalog No. H115925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H115925G 25 g
H1159500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H115925G Supplier TCI America Supplier No. H115925G
Only null left

Chemical Identifiers

CAS 35274-05-6
Molecular Formula C19H38O3
Molecular Weight (g/mol) 314.51
InChI Key WUKXMJCZWYUIRZ-UHFFFAOYSA-N
Synonym Cetyl Lactate, Hexadecyl 2-Hydroxypropionate, Lactic Acid Hexadecyl Ester
PubChem CID 37098
IUPAC Name hexadecyl 2-hydroxypropanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)C(C)O

Specifications

Melting Point 42°C
Color White
Quantity 25 g
Formula Weight 314.51
Percent Purity ≥75.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyl Lactate
EINECSNumber (9)-0951
TSCA Yes