missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanedioic Acid 97.0+%, TCI America™
$188.27 - $1979.63
Chemical Identifiers
| CAS | 505-54-4 |
|---|---|
| Molecular Formula | C16H30O4 |
| Molecular Weight (g/mol) | 286.412 |
| MDL Number | MFCD00002746 |
| InChI Key | QQHJDPROMQRDLA-UHFFFAOYSA-N |
| Synonym | thapsic acid, 1,14-tetradecanedicarboxylic acid, 1,16-hexadecanedioic acid, hexadecane-1,16-dioic acid, 1,14-tetradecane dicarboxylic acid, n-tetradecane-.omega.,.omega.'-dicarboxylic acid, dicarboxylic acid c16, 1,16-hexadecanedioate, thapsate, thaspic acid |
| PubChem CID | 10459 |
| ChEBI | CHEBI:73722 |
| IUPAC Name | hexadecanedioic acid |
| SMILES | C(CCCCCCCC(=O)O)CCCCCCC(=O)O |
Chemical Identifiers
| 505-54-4 | |
| 286.412 | |
| QQHJDPROMQRDLA-UHFFFAOYSA-N | |
| 10459 | |
| hexadecanedioic acid |
| C16H30O4 | |
| MFCD00002746 | |
| thapsic acid, 1,14-tetradecanedicarboxylic acid, 1,16-hexadecanedioic acid, hexadecane-1,16-dioic acid, 1,14-tetradecane dicarboxylic acid, n-tetradecane-.omega.,.omega.'-dicarboxylic acid, dicarboxylic acid c16, 1,16-hexadecanedioate, thapsate, thaspic acid | |
| CHEBI:73722 | |
| C(CCCCCCCC(=O)O)CCCCCCC(=O)O |
Specifications
| 505-54-4 | |
| White | |
| MFCD00002746 | |
| thapsic acid, 1,14-tetradecanedicarboxylic acid, 1,16-hexadecanedioic acid, hexadecane-1,16-dioic acid, 1,14-tetradecane dicarboxylic acid, n-tetradecane-.omega.,.omega.'-dicarboxylic acid, dicarboxylic acid c16, 1,16-hexadecanedioate, thapsate, thaspic acid | |
| C(CCCCCCCC(=O)O)CCCCCCC(=O)O | |
| 286.412 | |
| CHEBI:73722 | |
| ≥97.0% (GC,T) | |
| Hexadecanedioic Acid |
| 126°C | |
| C16H30O4 | |
| 1 g | |
| QQHJDPROMQRDLA-UHFFFAOYSA-N | |
| hexadecanedioic acid | |
| 10459 | |
| 286.41 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-3082
TSCA : Yes