missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane 98.0+%, TCI America™

Catalog No. H006625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H006625ML 25 mL
H0066100ML 100 mL
H0066500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H006625ML Supplier TCI America Supplier No. H006625ML

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.448
MDL Number MFCD00008998
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Color Colorless
Boiling Point 287°C
Quantity 25 mL
Formula Weight 226.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexadecane
EINECSNumber (2)-0010
RTECSNumber ML9200000
TSCA Yes