missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane, Reagents

Catalog No. 01337223
Change view
Click to view available options
Quantity:
100 mL
450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337223 100 mL
01337222 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337223 Supplier Labchem Inc Supplier No. C2213400100C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hexadecane, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.45
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym Cetane, n-Cetane, n-Hexadecane
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Boiling Range 287°C
Color Colorless
Boiling Point 287°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 0.773
Grade Reagent
Physical Form Liquid
Chemical Name or Material Hexadecane