missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane, Reagent, 99%, Spectrum™ Chemical

Catalog No. p-7157576
Change view
Click to view available options
Quantity:
100 mL
500 mL
18.1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603113 100 mL Amber Glass Bottle
18603114 500 mL Amber Glass Bottle
18603115 18.1 L Steel Pail
18603116 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603113 Supplier Spectrum Chemical Mfg Cor Supplier No. HE166100ML
Only null left

HE166, 544-76-3, C16H34

Spectrum™ Chemical Hexadecane, Reagent

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.45
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 99%
Grade Reagent