missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane, ASTM Grade, 99.0% min., MilliporeSigma™

Supplier:  Technidata HX02003

Catalog No. MHX02003

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hexadecane
544-76-3
0.77g/cm3 (20°C)
C16H34
250 g
DCAYPVUWAIABOU-UHFFFAOYSA-N
hexadecane
11006
226.44
Liquid
For Gas Chromatography
Colorless
99.0% min.
CH3(CH2)14CH3
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CCCCCCCCCCCCCCCC
226.448
CHEBI:45296
< - 0.1 hPa (20°C)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

SDS
Documents

Documents

Promotions

Promotions