missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Hexadecane

Catalog No. H67031L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H67031L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H67031L Supplier Sigma Aldrich Supplier No. H67031L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexadecane

Specifications

CAS 544-76-3
Melting Point 18°C
Density 0.773 g/mL at 25°C
Boiling Point 287°C
Molecular Formula C16H34
Refractive Index n20/D 1.434
Linear Formula CH3(CH2)14CH3
MDL Number MFCD00008998
Quantity 1 L
Synonym n-Hexadecane; Cetane
Molecular Weight (g/mol) 226.44 g/mol
Show More Show Less