missing translation for 'onlineSavingsMsg'
Learn More

Hexadecamethylcyclooctasiloxane 96.0+%, TCI America™

Catalog No. H12885ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12885ML 5 mL
H12881ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12885ML Supplier TCI America Supplier No. H12885ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 556-68-3
Molecular Formula C16H48O8Si8
Molecular Weight (g/mol) 593.232
InChI Key XKJMJYZFAWYREL-UHFFFAOYSA-N
PubChem CID 11170
IUPAC Name 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16-hexadecamethyl-1,3,5,7,9,11,13,15-octaoxa-2,4,6,8,10,12,14,16-octasilacyclohexadecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)C

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 mL
Formula Weight 593.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexadecamethylcyclooctasiloxane
EINECSNumber (7)-0475
TSCA No