missing translation for 'onlineSavingsMsg'
Learn More

Hexadecafluoro(1,3-dimethylcyclohexane) 80.0+%, TCI America™

Catalog No. p142025g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
P142025G 25 g
1 options

Catalog No. P142025G

Supplier: TCI America P142025G

Only null left

Chemical Identifiers

CAS 335-27-3
Molecular Formula C8F16
Molecular Weight (g/mol) 400.062
MDL Number MFCD00001469
InChI Key LOQGSOTUHASIHI-UHFFFAOYSA-N
Synonym perfluoro-1,3-dimethylcyclohexane, perfluoro 1,3-dimethylcyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl cyclohexane, decafluoro-1,3-bis trifluoromethyl cyclohexane, cyclohexane, perfluoro-1,3-dimethyl, cyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl, cyclohexane, decafluoro-1,3-bis trifluoromethyl, hexadecafluoro 1,3-dimethylcyclohexane, flutec pp3, acmc-1cjoy
PubChem CID 78975
IUPAC Name 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifications

Color Colorless
Quantity 25 g
Formula Weight 400.06
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexadecafluoro(1,3-dimethylcyclohexane)
EINECSNumber (3)-3247
RTECSNumber GV4990000
TSCA Yes