missing translation for 'onlineSavingsMsg'
Learn More

Hexacosanoic acid, 98%

CAS: 506-46-7 | C26H52O2 | 396.7 g/mol

Supplier:  Thermo Scientific Chemicals 258210010

 View more versions of this product

Catalog No. AC258210010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexacosanoic acid
506-46-7
White
250.0°C
Authentic
Glass bottle
1.4301
MFCD00002811
0.81
Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether and acetone
CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
396.7
CHEBI:31009
98%
98%
87.0°C to 89.0°C
0.8100g/mL
>110°C
98%
C26H52O2
CH3(CH2)24COOH
1 g
cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
XMHIUKTWLZUKEX-UHFFFAOYSA-N
hexacosanoic acid
10469
396.7
Shiny Fluffy Crystalline Powder

Chemical Identifiers

506-46-7
396.7
XMHIUKTWLZUKEX-UHFFFAOYSA-N
10469
hexacosanoic acid
C26H52O2
MFCD00002811
cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
CHEBI:31009
CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-040-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only