missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroplatinic(IV) Acid Solution (about 10%), (3.8% Pt) GR For analysis, MilliporeSigma™

$1720.76 - $1720.76

Chemical Identifiers

CAS 16941-12-1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1073410025
View Documents
Technidata
1073410025
25 mL Glass bottle
Each for $1,720.76
Only null left
 
Description

Description

For analysis
Specifications

Chemical Identifiers

16941-12-1

Specifications

16941-12-1
25 mL
UN 3264
Glass bottle
Hexachloroplatinic(IV) acid solution about
Liquid
10%
Odorless
Orange liquid
Videos
Safety and Handling

Safety and Handling

H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.

DOTInformation : DOT Class: 8, Packaging Group: III

SDS
Documents

Documents