missing translation for 'onlineSavingsMsg'
Learn More

Hexacarbonylchromium, 99%

Catalog No. p-4824743
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1306103 1 g
AA1306109 10 g
AA1306118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1306103 Supplier Thermo Scientific Chemicals Supplier No. 01306103
Only null left

CAS: 13007-92-6 | C6CrO6 | 220.056 g/mol

Hexacarbonylchromium is widely used as a catalyst for isomerization of alkenes. It is involved in the hydrogenation of 1,3-conjugated dienes and oxidation of allylic and benzylic positions. It acts as a precursor to prepare phenanthrenes, tricarbonyl(arene)chromium and Fischer carbine complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13007-92-6
Molecular Formula C6CrO6
Molecular Weight (g/mol) 220.056
MDL Number MFCD00010945
InChI Key KOTQLLUQLXWWDK-UHFFFAOYSA-N
Synonym chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g
PubChem CID 25589
ChEBI CHEBI:33031
IUPAC Name carbon monoxide;chromium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Specifications

Melting Point ∽152°C (decomposition)
Color White
Physical Form Crystals
Quantity 1 g
Assay Percent Range 99%
Solubility Information Soluble in ether,chloroform,tetrahydropyran and methylene chloride. Insoluble in water.
Formula Weight 220.06
Sensitivity Light sensitive
Vapor Pressure 0.35 hPa
Density 1.77
Chemical Name or Material Hexacarbonylchromium
Show More Show Less
Hazard Category H302+H312+H332-H317-H350i
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P333+P313-P363-P501c
DOTInformation DOT Class: 6.1, Packing Group: III
EINECSNumber 235-852-4
RTECSNumber GB5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only