missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexacarbonylchromium, 99%
Description
Hexacarbonylchromium is widely used as a catalyst for isomerization of alkenes. It is involved in the hydrogenation of 1,3-conjugated dienes and oxidation of allylic and benzylic positions. It acts as a precursor to prepare phenanthrenes, tricarbonyl(arene)chromium and Fischer carbine complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∽152°C (decomposition) |
| Color | White |
| Physical Form | Crystals |
| Quantity | 1 g |
| Assay Percent Range | 99% |
| Solubility Information | Soluble in ether,chloroform,tetrahydropyran and methylene chloride. Insoluble in water. |
| Formula Weight | 220.06 |
| Sensitivity | Light sensitive |
| Vapor Pressure | 0.35 hPa |
| Density | 1.77 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H317-H350i |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P333+P313-P363-P501c |
| DOTInformation | DOT Class: 6.1, Packing Group: III |
| EINECSNumber | 235-852-4 |
| RTECSNumber | GB5075000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only