missing translation for 'onlineSavingsMsg'
Learn More

Hexaammineruthenium(III) chloride, 98%

Catalog No. p-3735332
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363342500 250 mg Glass bottle
AC363340010 1 g Glass bottle
AC363340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC363342500 Supplier Thermo Scientific Chemicals Supplier No. 363342500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14282-91-8 | Cl3H18N6Ru | 309.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14282-91-8
Molecular Formula Cl3H18N6Ru
Molecular Weight (g/mol) 309.61
MDL Number MFCD00011478
InChI Key GBDZMMXUOBAJMN-UHFFFAOYSA-K
Synonym hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride
PubChem CID 159731
SMILES N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Color Yellow
Physical Form Crystalline Powder
Quantity 250 mg
Assay Percent Range 98%
Solubility Information Solubility in water: soluble
IUPAC Name azane;ruthenium(3+);trichloride
Formula Weight 309.61
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Hexaammineruthenium(III) chloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 238-176-8

RUO – Research Use Only