missing translation for 'onlineSavingsMsg'
Learn More

Hexaammineruthenium(II) chloride, 99.9%

Catalog No. p-7027265
Change view
Click to view available options
Quantity:
0.25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2388601 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2388601 Supplier Thermo Scientific Chemicals Supplier No. B2388601

CAS: 15305-72-3 | ClH12N6Ru | 232.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15305-72-3
Molecular Formula ClH12N6Ru
Molecular Weight (g/mol) 232.66
MDL Number MFCD00011477
InChI Key MHYZHXDLILNYTQ-UHFFFAOYSA-M
Synonym triton rw 30, azane;ruthenium 2+ ;dichloride, ruthenium iii chloride hexaammoniate, hexaammineruthenium ii chloride

Specifications

Melting Point >300°C
Odor Odorless
Quantity 0.25 g
Sensitivity Air sensitive
Formula Weight 274.16
Percent Purity 99.9%
Chemical Name or Material Hexaammineruthenium(II) chloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 239-342-2
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only