missing translation for 'onlineSavingsMsg'
Learn More

Het Anhydride 95.0+%, TCI America™

Catalog No. p-7130501
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H034925G 25 g
H0349500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H034925G Supplier TCI America Supplier No. H034925G
Only null left

Chemical Identifiers

CAS 115-27-5
Molecular Formula C9H2Cl6O3
Molecular Weight (g/mol) 370.812
MDL Number MFCD00080438
InChI Key FLBJFXNAEMSXGL-UHFFFAOYSA-N
Synonym chlorendic anhydride, het anhydride, chloran 542, 4,7-methanoisobenzofuran-1,3-dione, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro, 1,4,5,6,7,7-hexachloro-5-norbornene-2,3-dicarboxylic anhydride, chlorendic anhydride van, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, hexachloroendomethylene tetrahydrophthalic anhydride, hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-hexachloro
PubChem CID 8265
SMILES C12C(C(=O)OC1=O)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 238°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 370.81
Percent Purity ≥95.0% (T)
Chemical Name or Material Het Anhydride
EINECSNumber (4)-0621
RTECSNumber RB9080000
TSCA Yes
Recommended Storage Refrigerator