missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hesperetin 97.0+%, TCI America™
$38.51 - $100.11
Chemical Identifiers
| CAS | 520-33-2 |
|---|---|
| Molecular Formula | C16H14O6 |
| Molecular Weight (g/mol) | 302.28 |
| MDL Number | MFCD00075646 |
| InChI Key | AIONOLUJZLIMTK-AWEZNQCLSA-N |
| Synonym | hesperetin, hesperitin, 3',5,7-trihydroxy-4'-methoxyflavanone, hesperin, yso2, cyanidanon 4'-methyl ether 1626, 5,7,3'-trihydroxy-4'-methoxyflavanone, --hesperetin, prestwick_908, unii-q9q3d557f1 |
| PubChem CID | 72281 |
| ChEBI | CHEBI:28230 |
| IUPAC Name | (2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one |
| SMILES | COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1 |
Chemical Identifiers
| 520-33-2 | |
| 302.28 | |
| AIONOLUJZLIMTK-AWEZNQCLSA-N | |
| 72281 | |
| (2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one |
| C16H14O6 | |
| MFCD00075646 | |
| hesperetin, hesperitin, 3',5,7-trihydroxy-4'-methoxyflavanone, hesperin, yso2, cyanidanon 4'-methyl ether 1626, 5,7,3'-trihydroxy-4'-methoxyflavanone, --hesperetin, prestwick_908, unii-q9q3d557f1 | |
| CHEBI:28230 | |
| COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1 |
Specifications
| 520-33-2 | |
| White-Yellow | |
| MFCD00075646 | |
| hesperetin, hesperitin, 3',5,7-trihydroxy-4'-methoxyflavanone, hesperin, yso2, cyanidanon 4'-methyl ether 1626, 5,7,3'-trihydroxy-4'-methoxyflavanone, --hesperetin, prestwick_908, unii-q9q3d557f1 | |
| COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1 | |
| 302.28 | |
| CHEBI:28230 | |
| ≥97.0% (HPLC,T) | |
| Hesperetin |
| 232°C | |
| C16H14O6 | |
| 5 g | |
| AIONOLUJZLIMTK-AWEZNQCLSA-N | |
| (2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one | |
| 72281 | |
| 302.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No