missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Heratherm Light Chambers 750 L
Accurate and even light distribution with Long-lasting LED lighting for (ICH) light testing, plant growth and animal hatching — relative humidity and temperature are controlled to meet specified environment.
Specifications
| Alarm Types | Audible and Visible |
|---|---|
| Capacity (English) | 26.5 cu.ft |
| Capacity (English) Shelf | 66 lbs (stationary) |
| Capacity (Metric) Shelf | 30 kg (stationary) |
| Certifications/Compliance | UL, CE |
| Catalog Number | Mfr. No. | Door Style | For Use With (Application) | Includes | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Door Style | For Use With (Application) | Includes | Price | Quantity & Availability | |||
|
01672440
|
Thermo Scientific™
ELS750 |
Solid outer door with inner glass door | ICH light testing, plant growth, light testing, animal hatching | Light modules not included - ICH testing: 333979G01 / Standard: 333980G01 |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01672441
|
Thermo Scientific™
ELG750 |
Glass door | Plant growth, light testing, animal hatching | Light modules not included - for plant growth/testing/animal hatching: 333980G01 |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Accurate control of three parameters - Temperature above and below ambient, relative humidity and light - provide great environment in testing applications and for biological research with plants and animals. Choice of ICH module provides capability for efficient ICH light testing in Pharma.
- Uniform Temperature / relative humidity / light exposure for samples.
- Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
- LED light system provides specified light exposure on 1, 2 or 3 shelves; programmable to adapt light quality / intensity and on/off timing - e.g. for day-night cycles. Choose from two light modules (ordered separately):
- 1. ICH module, includes UV LED for ICH light testing;can only be used in ELS750 - model with solid outer door - for safety reasons
- 2. Testing module for plant growth / daylight testing / animal hatching
- Environmentally friendly water-efficient humidification system - with water tank or optional fix water connection for in-house purified water
- Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
- Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
- Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard (with Connect Edge solution)
- Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
- Simplified setup with no technician required; install using standard power cord.
- IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
- Inner glass door keeps environment stable, even when opening outer door. Exterior glass door also available (not for ICH module use - for safety reasons).
- Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
Specifications
| Audible and Visible | |
| 66 lbs (stationary) | |
| UL, CE | |
| 750 L | |
| Powder-coated Galvanized Steel | |
| Nema 5-20P | |
| Vertical Light Chamber | |
| 10 - 80% RH with light; 10 -90% RH w/o light | |
| Removable water tank | |
| Temperature, relative humidity, light intensity | |
| Stainless Steel (304/1.4301) | |
| 24 months | |
| 895 x 1080 x 1835 mm / 35.24 x 42.52 x 72.24 in | |
| 31.3 in. | |
| 100/240 V |
| 26.5 cu.ft | |
| 30 kg (stationary) | |
| USB, Thermo Cloud via OPC UA | |
| Stainless Steel (304/1.4301) | |
| 3 | |
| 10 to 90% | |
| 7 in. Touchscreen user interface | |
| 0.1% RH | |
| 2 access ports | |
| 0.46 m2 | |
| 0.1°C | |
| 0.05 - 1 MΩ | |
| 630 x 840 x 1450 mm / 24.8 x 33.07 x 57.09 in | |
| 100 - 240V, 50-60Hz, 1700W | |
| 1700 W |