missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Heratherm CO2 Chamber
Precisely control the essential parameters of your distinct cell growth or CO2 testing conditions - active Humidity controls the incubation environment with accuracy.
$56674.96 - $57766.06
Specifications
| Alarm Types | Audible and Visible |
|---|---|
| Capacity (English) Shelf | 66.1 lb. |
| Capacity (Metric) Shelf | 30 kg |
| Certifications/Compliance | UL, CE |
| Data Outputs | USB, Thermo Cloud via OPC UA |
| Catalog Number | Mfr. No. | Door Style | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Door Style | Price | Quantity | |||||
|
01672445PM
|
Thermo Scientific™
ECG750PM |
Glass Door |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01672444PM
|
Thermo Scientific™
ECS750PM |
Solid Outer, Glass Inner |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Uniform Temperature / CO2 / Relative Humidity throughout the chamber
- Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
- CO control system ensures stable CO2 environment, range of 0 to 20 Vol-%.
- Environmentally friendly water-efficient humidification system - with water tank or optional fixed water connection for in-house purified water
- Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
- Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
- Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard.
- Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
- Simplified setup with no technician required; install using standard power cord.
- IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
- Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
- Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
- Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option.
- Choose from a variety of country-specific sockets. (Ordered with separate order code)
Specifications
| Audible and Visible | |
| 30 kg | |
| USB, Thermo Cloud via OPC UA | |
| Stainless Steel (304/1.4301) | |
| 3 | |
| 10 to 90% | |
| Touchscreen | |
| 10 to 90% RH | |
| Removable water tank | |
| 2 Access Ports | |
| 4.9 ft.2 | |
| Stainless Steel (304/1.4301) | |
| 24 Months | |
| 0.05 to 1 MOhm | |
| 24.8 x 33.07 x 57.09 in. (63 x 84 x 145 cm) | |
| 79.5 cm | |
| 390 kg | |
| 100/115/220/230 V |
| 66.1 lb. | |
| UL, CE | |
| 750 L | |
| Powder-coated Galvanized Steel | |
| Nema 5-20P | |
| Growth Chamber | |
| Cell growth, testing | |
| 0.1% RH | |
| 4 | |
| Temperature, Relative Humidity, CO2 Ramping | |
| 0.46 m2 | |
| Microprocessor | |
| <0.8 L/day | |
| 35.24 x 42.52 x 72.24 in. (89.5 x 108 x 183.5 cm) | |
| 31.3 in. | |
| 859 lb. | |
| 100/240 V, 50/60 Hz, 1400 W | |
| 1400 W |