missing translation for 'onlineSavingsMsg'
Learn More

Heptyl Chloroformate 98.0+%, TCI America™

Catalog No. p-7124576
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07225ML 5 mL
C072225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07225ML Supplier TCI America Supplier No. C07225ML

Chemical Identifiers

CAS 33758-34-8
Molecular Formula C8H15ClO2
Molecular Weight (g/mol) 178.656
MDL Number MFCD00059484
InChI Key SATRZZYUXUGZIE-UHFFFAOYSA-N
Synonym Chloroformic Acid Heptyl Ester
PubChem CID 118547
IUPAC Name heptyl carbonochloridate
SMILES CCCCCCCOC(=O)Cl

Specifications

Color Colorless
Boiling Point 71°C
Quantity 5 mL
UN Number 3277
Formula Weight 178.66
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Heptyl Chloroformate