missing translation for 'onlineSavingsMsg'
Learn More

Heptanoyl chloride, 99%

Catalog No. AC164261000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164261000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC164261000 Supplier Thermo Scientific Chemicals Supplier No. 164261000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2528-61-2 | C7H13ClO | 148.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2528-61-2
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD00000763
InChI Key UCVODTZQZHMTPN-UHFFFAOYSA-N
Synonym enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
PubChem CID 17313
IUPAC Name heptanoyl chloride
SMILES CCCCCCC(Cl)=O

Specifications

Melting Point -84°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 173°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4295 to 1.4315
Linear Formula CH3(CH2)5COCl
Quantity 100 g
Beilstein 02, 340
Specific Gravity 0.96
Solubility Information Solubility in water: hydrolysis. Other solubilities: slightly soluble in carbontetrachloride,soluble in ethylether,ligroin
Formula Weight 148.63
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Heptanoyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 219-775-3
TSCA TSCA

RUO – Research Use Only