missing translation for 'onlineSavingsMsg'
Learn More

Heptanoyl chloride, 99%

Catalog No. AAL0331518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0331518 50 g
AAL0331530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0331518 Supplier Thermo Scientific Chemicals Supplier No. L0331518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2528-61-2 | C7H13ClO | 148.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2528-61-2
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD00000763
InChI Key UCVODTZQZHMTPN-UHFFFAOYSA-N
Synonym enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
PubChem CID 17313
IUPAC Name heptanoyl chloride
SMILES CCCCCCC(Cl)=O

Specifications

Melting Point -84°C
Density 0.96
Boiling Point 172°C to 174°C
Flash Point 58°C (136°F)
Odor Acrid
Refractive Index 1.431
Linear Formula CH3(CH2)5COCl
Quantity 50 g
UN Number UN2920
Beilstein 605636
Sensitivity Moisture sensitive
Formula Weight 148.63
Percent Purity 99%
Chemical Name or Material Heptanoyl chloride
Show More Show Less
Hazard Category H227-H290-H314-H330
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P234-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 219-775-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only