missing translation for 'onlineSavingsMsg'
Learn More

Heptanenitrile 98.0+%, TCI America™

Catalog No. H088325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H088325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H088325ML Supplier TCI America Supplier No. H088325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-08-3
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.19
MDL Number MFCD00039503
InChI Key SDAXRHHPNYTELL-UHFFFAOYSA-N
Synonym heptanonitrile, hexyl cyanide, enanthonitrile, 1-cyanohexane, 1-heptanonitrile, heptane-1-nitrile, n-heptanenitrile, unii-ua4z3qt98a, ua4z3qt98a, heptane nitrile
PubChem CID 12372
IUPAC Name heptanenitrile
SMILES CCCCCCC#N

Specifications

Color Colorless
Boiling Point 183°C
Quantity 25 mL
UN Number 1993
Formula Weight 111.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptanenitrile