missing translation for 'onlineSavingsMsg'
Learn More

Heptanal 95.0+%, TCI America™

Catalog No. p-7130293
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H002525ML 25 mL
H0025500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H002525ML Supplier TCI America Supplier No. H002525ML
Only null left

Chemical Identifiers

CAS 111-71-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00007028
InChI Key FXHGMKSSBGDXIY-UHFFFAOYSA-N
Synonym heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
PubChem CID 8130
ChEBI CHEBI:34787
IUPAC Name heptanal
SMILES CCCCCCC=O

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
UN Number 3056
Formula Weight 114.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptanal
EINECSNumber (2)-0217&(2)-0494
RTECSNumber MI6900000
TSCA Yes