missing translation for 'onlineSavingsMsg'
Learn More

Heptanal 95.0+%, TCI America™

Catalog No. h002525ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
H002525ML 25 mL
H0025500ML 500 mL
2 options

Catalog No. H002525ML

Supplier: TCI America H002525ML

Only null left

Chemical Identifiers

CAS 111-71-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00007028
InChI Key FXHGMKSSBGDXIY-UHFFFAOYSA-N
Synonym heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
PubChem CID 8130
ChEBI CHEBI:34787
IUPAC Name heptanal
SMILES CCCCCCC=O

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
UN Number 3056
Formula Weight 114.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptanal
EINECSNumber (2)-0217&(2)-0494
RTECSNumber MI6900000
TSCA Yes