missing translation for 'onlineSavingsMsg'
Learn More

Heptanal, 97%

Catalog No. aab23830ap
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
1000 mL
250 mL
500 mL
Catalog No. Quantity
AAB23830AP 500 mL
AAB23830AE 100 mL
AAB23830AK 250 mL
AAB23830AU 1000 mL
4 options

Catalog No. AAB23830AP

Supplier: Thermo Scientific Chemicals B23830AP

Only null left

CAS: 111-71-7 | C7H14O | 114.19 g/mol

Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

Solubility
Soluble in water 1.3 g/L.

Notes
Hygroscopic. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 111-71-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00007028
InChI Key FXHGMKSSBGDXIY-UHFFFAOYSA-N
Synonym heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
PubChem CID 8130
ChEBI CHEBI:34787
IUPAC Name heptanal
SMILES CCCCCCC=O

Specifications

Melting Point -43°C
Density 0.85
Boiling Point 152°C to 154°C
Flash Point 35°C (95°F)
Odor Unpleasant
Assay Percent Range 0.97
Refractive Index 1.4125
Quantity 500 mL
UN Number UN3056
Beilstein 1560236
Sensitivity Air Sensitive
Merck Index 14,4658
Solubility Information Soluble in water 1.3g/L.
Formula Weight 114.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Heptanal
Show More Show Less
Hazard Category H226-H315
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-HEPTALDEHYDE
EINECSNumber 203-898-4
RTECSNumber MI6900000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only