missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptakis(2,6-di-o-methyl)-β-cyclodextrin, MP Biomedicals™
Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a cholesterol sequestering agent
$145.15 - $458.28
Chemical Identifiers
| CAS | 51166-71-3 |
|---|---|
| Molecular Formula | C56H98O35 |
| Molecular Weight (g/mol) | 1331.365 |
| MDL Number | MFCD00011616 |
| InChI Key | QGKBSGBYSPTPKJ-UZMKXNTCSA-N |
| Synonym | 2,6-di-o-methyl-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dimethyl, dimethyl, 2,6-dme-beta-cyclodextrin, 2,6-dme-beta-cyclodextrin |
| PubChem CID | 10171019 |
| SMILES | COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15732001
|
MP Biomedicals Inc
0215732001 |
1 g |
N/A
|
|
|||||
|
ICN15732005
|
MP Biomedicals Inc
0215732005 |
5 g |
N/A
|
|
|||||
Chemical Identifiers
| 51166-71-3 | |
| 1331.365 | |
| QGKBSGBYSPTPKJ-UZMKXNTCSA-N | |
| 10171019 |
| C56H98O35 | |
| MFCD00011616 | |
| 2,6-di-o-methyl-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dimethyl, dimethyl, 2,6-dme-beta-cyclodextrin, 2,6-dme-beta-cyclodextrin | |
| COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O |
Specifications
| 51166-71-3 | |
| C56H98O35 | |
| 1 g | |
| QGKBSGBYSPTPKJ-UZMKXNTCSA-N | |
| 1331.365 | |
| 1331.4 | |
| Heptakis(2, 6-di-o-methyl)-β-cyclodextrin |
| White | |
| MFCD00011616 | |
| 2,6-di-o-methyl-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dimethyl, dimethyl, 2,6-dme-beta-cyclodextrin, 2,6-dme-beta-cyclodextrin | |
| COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O | |
| 10171019 | |
| Powder |