missing translation for 'onlineSavingsMsg'
Learn More

Heptafluoropropyl Bromide 98.0+%, TCI America™

Catalog No. h06895g
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
H06895G 5 g
1 options

Catalog No. H06895G

Supplier: TCI America H06895G

Only null left

Chemical Identifiers

CAS 422-85-5
Molecular Formula C3BrF7
Molecular Weight (g/mol) 248.926
MDL Number MFCD00039238
InChI Key LANNRYWUUQMNPF-UHFFFAOYSA-N
Synonym Perfluoropropyl Bromide
PubChem CID 67916
IUPAC Name 1-bromo-1,1,2,2,3,3,3-heptafluoropropane
SMILES C(C(F)(F)F)(C(F)(F)Br)(F)F

Specifications

Boiling Point 12°C
Quantity 5 g
Formula Weight 248.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptafluoropropyl Bromide