missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric anhydride, 98%

Catalog No. AC169260250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169260250 25 g Glass bottle
AC169261000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169260250 Supplier Thermo Scientific Chemicals Supplier No. 169260250

CAS: 336-59-4 | C8F14O3 | 410.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent

Chemical Identifiers

CAS 336-59-4
Molecular Formula C8F14O3
Molecular Weight (g/mol) 410.06
MDL Number MFCD00000432
InChI Key UFFSXJKVKBQEHC-UHFFFAOYSA-N
Synonym heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization
PubChem CID 67643
ChEBI CHEBI:39424
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point -43°C
Density 1.6500g/mL
Boiling Point 108°C to 111°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CF3CF2CF2CO)2O
Quantity 25 g
Specific Gravity 1.65
Solubility Information Solubility in water: reacts with water forming heptafluorobutyric acid
Formula Weight 410.05
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Heptafluorobutyric anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 206-410-8

RUO – Research Use Only