missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric acid, For ion chromatography, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020371
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020371 5 mL
111018870 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020371 Supplier Merck Emd Millipore Supplier No. 524115MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00004171
Synonym Edman Reagent No. 3; HFBA; Perfluorobutyric acid

Specifications

CAS 375-22-4
Density 1.645 g/mL (at 25°C)
Boiling Point 120°C (755 mmHg)
Packaging Glass Bottle
Linear Formula CF3 CF2 CF2 COOH
Quantity 5 mL
Vapor Density 7 (vs. air)
Vapor Pressure ˜10 mmHg (25°C)
Formula Weight 214.04
Percent Purity ≥99.5% (GC)
Grade For ion chromatography
Show More Show Less
ShelfLife Limited shelf life, expiry date on the label
RTECSNumber ET4025000