missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptaethylene Glycol Monomethyl Ether 97.0+%, TCI America™
$201.32 - $695.29
Chemical Identifiers
| CAS | 4437-01-8 |
|---|---|
| Molecular Formula | C15H32O8 |
| Molecular Weight (g/mol) | 340.41 |
| MDL Number | MFCD06201006 |
| InChI Key | AGWKUHGLWHMYTG-UHFFFAOYSA-N |
| Synonym | mPEG7-Alcohol |
| PubChem CID | 526555 |
| IUPAC Name | 2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol |
| SMILES | COCCOCCOCCOCCOCCOCCOCCO |
Chemical Identifiers
| 4437-01-8 | |
| 340.41 | |
| AGWKUHGLWHMYTG-UHFFFAOYSA-N | |
| 526555 | |
| COCCOCCOCCOCCOCCOCCOCCO |
| C15H32O8 | |
| MFCD06201006 | |
| mPEG7-Alcohol | |
| 2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol |
Specifications
| 4437-01-8 | |
| C15H32O8 | |
| 1 g | |
| AGWKUHGLWHMYTG-UHFFFAOYSA-N | |
| 2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol | |
| 526555 | |
| ≥97.0% (GC) | |
| Heptaethylene Glycol Monomethyl Ether |
| Yellow | |
| MFCD06201006 | |
| mPEG7-Alcohol | |
| COCCOCCOCCOCCOCCOCCOCCO | |
| 340.41 | |
| 340.41 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-0097
TSCA : No