missing translation for 'onlineSavingsMsg'
Learn More

Heptadecyltrimethylammonium Bromide 98.0+%, TCI America™

Catalog No. H13075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13075G Supplier TCI America Supplier No. H13075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21424-24-8
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.483
InChI Key TTXDNWCDEIIMDP-UHFFFAOYSA-M
Synonym Trimethylheptadecylammonium Bromide
PubChem CID 10045219
IUPAC Name heptadecyl(trimethyl)azanium;bromide
SMILES CCCCCCCCCCCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Color White
Quantity 5 g
Formula Weight 378.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecyltrimethylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA No