missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecanoyl Chloride 98.0+%, TCI America™
Supplier: TCI America H078225ML
Specifications
| Heptadecanoyl Chloride | |
| Yellow | |
| C17H33ClO | |
| 25 mL | |
| Margaroyl Chloride | |
| CCCCCCCCCCCCCCCCC(=O)Cl | |
| 288.9 | |
| 288.90 | |
| Liquid |
| 40480-10-2 | |
| 176°C | |
| MFCD00000743 | |
| 3265 | |
| ICDQUAGMQCUEMY-UHFFFAOYSA-N | |
| heptadecanoyl chloride | |
| 3516438 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 40480-10-2 | |
| 288.9 | |
| ICDQUAGMQCUEMY-UHFFFAOYSA-N | |
| 3516438 | |
| CCCCCCCCCCCCCCCCC(=O)Cl |
| C17H33ClO | |
| MFCD00000743 | |
| Margaroyl Chloride | |
| heptadecanoyl chloride |
Safety and Handling
TSCA : No