missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecanoic acid, 98%
Description
Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.
Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly). Insoluble in water.
Notes
Store at 4°C. Store away from oxidizing agents.
Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.
Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly). Insoluble in water.
Notes
Store at 4°C. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 59°C to 63°C |
| Density | 0.853 |
| Boiling Point | 227°C (100 mmHg) |
| Linear Formula | CH3(CH2)15CO2H |
| Quantity | 5 g |
| Beilstein | 1781004 |
| Merck Index | 14,5751 |
| Solubility Information | Soluble in Chloroform (Slightly),Methanol (Slightly). Insoluble in water. |
| Formula Weight | 270.46 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 208-027-1 |
| RTECSNumber | MI3850000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only