missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic acid, 98%

Catalog No. AC329740050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329740050 5 g Glass bottle
AC329740010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329740050 Supplier Thermo Scientific Chemicals Supplier No. 329740050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 506-12-7 | C17H34O2 | 270.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 506-12-7
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.44
MDL Number MFCD00002751
InChI Key KEMQGTRYUADPNZ-UHFFFAOYSA-N
Synonym margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
PubChem CID 10465
ChEBI CHEBI:32365
IUPAC Name heptadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 59°C to 61°C
Color White
Boiling Point 364°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)15CO2H
Quantity 5 g
Beilstein 02, 376
Merck Index 15, 5816
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and acetone
Formula Weight 270.44
Percent Purity 98%
Physical Form Shiny Crystalline Powder or Flakes
Chemical Name or Material Heptadecanoic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-027-1
RTECSNumber MI3850000
TSCA TSCA

RUO – Research Use Only