missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanal 97.0+%, TCI America™

Catalog No. H12951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12951G Supplier TCI America Supplier No. H12951G
Only null left

Chemical Identifiers

CAS 629-90-3
Molecular Formula C17H34O
Molecular Weight (g/mol) 254.458
MDL Number MFCD04034658
InChI Key PIYDVAYKYBWPPY-UHFFFAOYSA-N
Synonym Heptadecanaldehyde, Heptadecyl Aldehyde
PubChem CID 71552
IUPAC Name heptadecanal
SMILES CCCCCCCCCCCCCCCCC=O

Specifications

Melting Point 34°C
Color White
Boiling Point 318°C
Quantity 1 g
Formula Weight 254.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecanal
EINECSNumber (2)-0494
RTECSNumber MI3210000
TSCA No
Recommended Storage Refrigerator