missing translation for 'onlineSavingsMsg'
Learn More

Heptadecan-9-amine 96.0+%, TCI America™

Catalog No. p-7131138
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16401ML 1 mL
H16405ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16401ML Supplier TCI America Supplier No. H16401ML
Only null left

Chemical Identifiers

CAS 3241-20-1
Molecular Formula C17H37N
Synonym 1-Octylnonylamine, 9-Aminoheptadecane

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 mL
UN Number 2735
Formula Weight 255.49
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Heptadecan-9-amine
EINECSNumber (2)-0133&(2)-0176
TSCA No