missing translation for 'onlineSavingsMsg'
Learn More

Heptacosane 97.0+%, TCI America™

Catalog No. p-7130286
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00171G 1 g
H001710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H00171G Supplier TCI America Supplier No. H00171G
Only null left

Chemical Identifiers

CAS 593-49-7
Molecular Formula C27H56
Molecular Weight (g/mol) 380.745
MDL Number MFCD00009862
InChI Key BJQWYEJQWHSSCJ-UHFFFAOYSA-N
Synonym n-heptacosane, unii-vp371w2gjs, vp371w2gjs, hexacosane, methyl, acmc-1aog8, qspl 047, qspl 073, heptacosane, analytical standard, ch3-ch2 25-ch3, heptacosane gc
PubChem CID 11636
ChEBI CHEBI:32941
IUPAC Name heptacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 59°C
Color White
Boiling Point 270°C
Quantity 1 g
Formula Weight 380.75
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heptacosane
EINECSNumber (2)-0010
TSCA No