missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES sodium salt, 99%

Catalog No. AAA1651614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1651614 25 g
AAA1651622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1651614 Supplier Thermo Scientific Chemicals Supplier No. A1651614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol

HEPES [N-(2-hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES [N-(2-hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

Solubility
It is soluble in water.

Notes
Stable under ordinary conditions. Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
IUPAC Name sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
Formula Weight 260.28
Percent Purity 99%
Chemical Name or Material HEPES sodium salt
EINECSNumber 278-169-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only