missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepatoZYME-SFM
GREENER_CHOICE

Catalog No. 17705021
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17705021 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17705021 Supplier Gibco™ Supplier No. 17705021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HepatoZYME-SFM

Hepatozyme-SFM is a serum-free medium for the long-term maintenance of hepatocyte phenotypic expression, including active and inducible forms of cytochromes P450 and active phase II enzymes.

Contains no phenol red.

Specifications

Cell Type Hepatocyte Cells
Classification Serum-free
Form Liquid
Product Type Hepatocyte Serum Free Medium (SFM)
Serum Level Serum-free
Without Additives No Glutamine, No Phenol Red
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture

For Research Use Only. Not for use in diagnostic procedures.